Skip to Content
MilliporeSigma

ENAH3035E40D

Enamine

carbamimidamidomethanimidamide, sulfuric acid

Synonym(s):

carbamimidamidomethanimidamide; sulfuric acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C2H9O4N5S1
CAS Number:
Molecular Weight:
199.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C2H7N5.H2O4S/c3-1(4)7-2(5)6;1-5(2,3)4/h(H7,3,4,5,6,7);(H2,1,2,3,4)

Inchi Key

MIWVBVPUEOHJFV-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95

Salt Data

O=S(=O)(O)O

Storage temp.

RT

Country of Origin

UA

Physical Form

powder



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library