Skip to Content
MilliporeSigma

ENAH304994F2

Enamine

methyl 3'-methylspiro[bicyclo[4.2.0]octane-7,2'-oxirane]-3'-carboxylate

Synonym(s):

methyl 3'-methylspiro[bicyclo[4.2.0]octane-7,2'-oxirane]-3'-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H18O3/c1-11(10(13)14-2)12(15-11)7-8-5-3-4-6-9(8)12/h8-9H,3-7H2,1-2H3

Inchi Key

PFFJCDANZPITOI-UHFFFAOYSA-N

Country of Origin

UA





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library