Skip to Content
MilliporeSigma

ENAH304C0586

Enamine

hexahydro-1H-spiro[indolizine-7,2'-oxirane]

Synonym(s):

hexahydro-1H-spiro[indolizine-7,2'-oxirane]

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
153.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H15NO/c1-2-8-6-9(7-11-9)3-5-10(8)4-1/h8H,1-7H2

Inchi Key

UJJWKURFLUZITI-UHFFFAOYSA-N

Country of Origin

UA





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library