Skip to Content
MilliporeSigma

ENAH304DF773

Enamine

3,3-diethyl-2,9-dioxa-4-azaspiro[5.5]undecane

Synonym(s):

3,3-diethyl-2,9-dioxa-4-azaspiro[5.5]undecane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H23NO2/c1-3-12(4-2)13-9-11(10-15-12)5-7-14-8-6-11/h13H,3-10H2,1-2H3

Inchi Key

NABHDUFXPFHXQR-UHFFFAOYSA-N

Country of Origin

UA





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library