Skip to Content
MilliporeSigma

ENAH9AD0F6A8

Enamine

rac-(1R,6R)-spiro[bicyclo[4.2.0]octane-7,2'-oxirane]-3'-carbonitrile

Synonym(s):

(1R,6R)-spiro[bicyclo[4.2.0]octane-7,2'-oxirane]-3'-carbonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H13NO/c11-6-9-10(12-9)5-7-3-1-2-4-8(7)10/h7-9H,1-5H2/t7-,8-,9?,10?/m1/s1

Inchi Key

RRJDFSWILIGTHJ-LGUIWLBCSA-N

Country of Origin

UA





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library