Skip to Content
MilliporeSigma

ENAH9AD0F6A9

Enamine

rac-ethyl (1R,6R)-spiro[bicyclo[4.2.0]octane-7,2'-oxirane]-3'-carboxylate

Synonym(s):

ethyl (1R,6R)-spiro[bicyclo[4.2.0]octane-7,2'-oxirane]-3'-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H18O3/c1-2-14-11(13)10-12(15-10)7-8-5-3-4-6-9(8)12/h8-10H,2-7H2,1H3/t8-,9-,10?,12?/m1/s1

Inchi Key

ZOLUMPWWXYSKES-XTQFYOIMSA-N

Country of Origin

UA





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library