Skip to Content
MilliporeSigma

ENAH9AD1287D

Enamine

rac-methyl (1R,6R)-spiro[bicyclo[4.2.0]octane-7,2'-oxirane]-3'-carboxylate

Synonym(s):

methyl (1R,6R)-spiro[bicyclo[4.2.0]octane-7,2'-oxirane]-3'-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H16O3/c1-13-10(12)9-11(14-9)6-7-4-2-3-5-8(7)11/h7-9H,2-6H2,1H3/t7-,8-,9?,11?/m1/s1

Inchi Key

RSXITAMLTXISLY-BFWVQVCMSA-N

Country of Origin

UA





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library