Skip to Content
MilliporeSigma

EXBR008

Exreprotein

Exreprotein™ EXREsolve™ Precast 4-20% PAGE Gel

12 Wells, 1 Kit

Shipped & Sold By Exreprotein LLC

A Trusted Marketplace Seller

Synonym(s):

Electrophoresis gel, Polyacrylamide gel electrophoresis

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


technique(s)

electrophoresis

manufacturer/tradename

Exreprotein

General description

Key Features:

  • Convenient and time-saving precast gels
  • Sharp bands
  • Batch-to-batch consistency
  • Instant MOPS buffer included

Legal Information

Exreprotein and EXREsolve are trademarks of Exreprotein, LLC.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library