Skip to Content
MilliporeSigma

EXHK027

Exreprotein

Exreprotein™ Human IFN-gamma Precoated ELISA Kit

1 Kit

Shipped & Sold By Exreprotein LLC

A Trusted Marketplace Seller

Synonym(s):

IFNg, hIFNg, hIFN-g, hIFN-gamma

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


technique(s)

ELISA

detection method

colorimetric

species reactivity

human

manufacturer/tradename

Exreprotein

General description

Key Features:

  • Assay Range: 12.5-400 pg/mL
  • Sensitivity: 5 pg/mL
  • Sample Types: Serum, Plasma, Cell Culture Supernates

Legal Information

Exreprotein is a trademark of Exreprotein, LLC.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library