MilliporeSigma
FITE-LAV-3G
Fischer Technical Company
Fischer Technical Company High Vacuum Pump
3 CFM With Gauge 110 V
Shipped & Sold By Fischer Technical Company
A Trusted Marketplace Seller
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
Fischer Technical Company
General description
These rotary style, 2 stage direct drive Pumps are ideal for use in virtually all High Vacuum applications.
  • Our Pumps are tested to 15 microns Hg but are suitable for all applications requirIng a vacuum under 500 microns Hg.
  • The Pumps use High Vacuum Pump Oil (one quart included) and are designed to run on a closed and dry system.
  • Regular oil changes and proper maintenance will ensure these Pumps will last for years in your laboratory.
  • Available with or without a mounted Bourdon Tube Gauge.
  • Available in four different flow rates: 3 CFM, 5 CFM, 7 CFM, and 10 CFM.
  • Available in either 110 V or 220 V.
Legal Information
Fischer Technical Company and it's logo is a trademark of Fischer Technical Company.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)