MilliporeSigma
FITE-OIL-012
Fischer Technical Company
Fischer Technical Company High Vacuum Pump Oil
One Quart Pack/12
Shipped & Sold By Fischer Technical Company
A Trusted Marketplace Seller
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
Fischer Technical Company
General description
High Vacuum Pump Oil. For use in any Fischer Technical LAV-Series vacuum pumps or other rotary vane pumps requiring high vacuum oil.
  • Our oil is extremely pure.
  • It is hydro processed, meaning it goes through a series of catalytic steps rendering the oil extremely refined, more viscous, and more stable.
  • The result is clear mineral oil which will alert you to contamination sooner as it becomes cloudy or milky.
  • In extreme temperatures, the oil will be more resistant to contaminants.
Legal Information
Fischer Technical Company and it's logo is a trademark of Fischer Technical Company.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)