MilliporeSigma
FITE-PFKIT5
Fischer Technical Company
Fischer Technical Company Safety Pipette Filler Assortment
Assortment Kit/5
Shipped & Sold By Fischer Technical Company
A Trusted Marketplace Seller
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
Fischer Technical Company
General description
Includes five natural rubber three valve pipette fillers, one each in Red, Black, Blue, Green and Multicolored pipette filler.
  • Accommodates pipettes up to 100ml (bulbs draw 50ml at a time, for volumes larger than 50ml user will have to repeat the liquid draw process).
Legal Information
Fischer Technical Company and it's logo is a trademark of Fischer Technical Company.
Choose from one of the most recent versions:
Certificates of Analysis (COA)