MilliporeSigma
PKE-50CB-NS
Fluotics
Fluotics Revvity Varispan P50
50 uL, conductive, non-sterile, 96 format
Shipped & Sold By Fluotics
A Trusted Marketplace Seller
Synonym(s):
Disposable pipette tips, conductive, non-sterile, Janus
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
binding type
non-treated surface
volume
50 μL
manufacturer/tradename
Fluotics
General description
Fluotics produces its PerkinElmer (Revvity) Janus compatible P50 tips from the highest grade virgin polypropylene. We use ultra polishing techniques to ensure low liquid retention without the use of releasing agents or other additives. When using Fluotics automation tips a separate labware definition is unnecessary and existing automation control software protocols also do not need to be modified.
  • Working volume: 2.00 - 50.00 uL
  • Tip format: non-sterile
  • Well format: 96, 384 plate accessible
  • Head: Janus Varispan
  • Tip material: conductive polypropylene
  • Tip box material: polypropylene
  • Case quantity: 96 tips per rack and 50 racks per case
Still not finding the right product?
Explore all of our products under Fluotics Revvity Varispan P50
Choose from one of the most recent versions:
Certificates of Analysis (COA)