MilliporeSigma
100A-O620
Glas-Col®
Glas-Col® Heating Mantle
4 Liter
Shipped & Sold By Glas-Col
A Trusted Marketplace Seller
Synonym(s):
heating mantle bottom, heating mantle
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
AC/DC
115
feature
CSA approved
heater capacity
710W
manufacturer/tradename
Glas-Col
General description
Fabric mantle for 4 liter griffin beaker.
  • 710 Watts
  • 115 Volts
  • Max Temperature 400oC
  • CSA Approved
  • A controller is required for operating mantles
Legal Information
Glas-Col is a registered trademark of Glas-Col LLC
Still not finding the right product?
Explore all of our products under Glas-Col® Heating Mantle
Choose from one of the most recent versions:
Certificates of Analysis (COA)