Skip to Content
MilliporeSigma

200A-HS3000A1

Glas-Col®

Glas-Col® Single Hose Fume Extractor

Single Hose Scavenger Fume Extractor

Shipped & Sold By Glas-Col

A Trusted Marketplace Seller

Synonym(s):

Fume Vacuum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


manufacturer/tradename

Glas-Col

General description

Single 5' Hose Portable Fume Vacuum

Scavenger Series portable fume/particle extractors are an inexpensive solution to all small scale, yet potentially hazardous, fume and particle problems. These powerful yet quiet, fully portable systems are ideal for use in work spaces that are too small for conventional fume hoods.

  • Work Space: Single Hose
  • Dimensions: Blower 16''H x 12.5''D x 23.5''W
  • Noise Level: A weighted noise level 68 dB max. at 1 meter.
  • Filter Size: 10" x 17"
  • Capacity: Pre-Filter/HEPA/Carbon
  • Volts: 120 Volts
  • Watts: 100
  • Includes: One 5" diameter hose and one tapered collector scoop

Legal Information

Glas-Col is a registered trademark of Glas-Col LLC




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library