MilliporeSigma
GLOBE-140105
Globe Scientific
Globe Scientific Reagent Reservoirs
Reagent Reservoir, 10mL, Disposable, White, PS, Graduated, Sterile, 5/Pack, 200/Unit-CS/200
Shipped & Sold By Globe Scientific LLC
A Trusted Marketplace Seller
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
Globe Scientific
General description
These reagent reservoirs are suitable for virtually all of your lab′s sample preparation and aliquoting needs. Design enhancements include pour spouts at each corner, a rigid sidewall design and graduations for easy volume reference. Sterile variants are irradiated to guarantee a sterility assurance level (SAL) of 10-6. Available volumes of 10mL, 25mL and 50mL ensure there is a volume and packaging option to fit almost every application. Individual, bulk and five-pack options are available.
  • Pour spouts at every corner
  • Rigid sidewall design
  • Wide, stable base
  • Lot traceability
  • Compatible with multi-channel pipettors
  • Bulk sterile & sterile options available
  • Molded graduations for easy volume reference
  • Easily stacked using minimal storage with easy separation
  • Available in 10mL, 25mL & 50mL volumes
  • Design allows for high level of solution recovery
  • DNase, RNase, pyrogen, animal materials and latex free
  • Produced from virgin medical grade polystyrene
Our universal Reagent Reservoir (Item # 140100) allows for one reservoir to work with single, eight & twelve channel pipettors. Providing one 50mL single reagent reservoir, one reservoir with 8 individual 5ml reagent channels & one reservoir with 12 individual 5mL reagent channels. This universal reagent reservoir is produced from polypropylene and is autoclavable for re-use.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)