MilliporeSigma
GLOBE-8100500
Globe Scientific
Globe Scientific Globe Glass Media Bottles
Bottle, Media, Globe Glass, 500mL, GL45 Screw Cap, Dual Graduations, 10/Box
Shipped & Sold By Globe Scientific LLC
A Trusted Marketplace Seller
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
feature
with cap
material
Bottle: borosilicate glass
manufacturer/tradename
Globe Scientific
General description
Globe Glass media bottles are designed with a wide mouth for ease of filling, dispensing and pipetting. A drip free pour ring and autoclavable polypropylene (PP) screw cap enable sterilizations of media and reagents. Globe Glass media bottles are available in sizes 50mL to 10000mL.
  • ASTM E438, Type I, Class A, Borosilicate 3.3 Glass
  • GL32 polypropylene (PP) screw cap on size 50mL
  • GL45 polypropylene (PP) screw caps on sizes 100mL to 10,000mL
  • Pouring ring for drip free liquid transfer
  • High resistance to chemicals
  • Dual scale metric graduations
  • Durable white enamel printing
  • Large white marking area
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)