MilliporeSigma
GLOBE-8400500
Globe Scientific
Globe Scientific Globe Glass Erlenmeyer Flasks - Narrow Mouth
Flask, Erlenmeyer, Globe Glass, 500mL, Narrow Mouth, Dual Graduations, ASTM E1404, 6/Box
Shipped & Sold By Globe Scientific LLC
A Trusted Marketplace Seller
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
Globe Scientific
General description
Globe Glass™ Erlenmeyer flasks meet ASTM E1404 specifications for narrow mouth, wide mouth, and screw cap options and are available in a wide range of sizes. Wide mouth conical flasks allow easy access to contents, while screw cap conical flasks include a GL45 cap with a PTFE liner for a leak-resistant closure. Globe Glass narrow and wide mouth Erlenmeyer flasks are designed with reinforced, beaded rims for strength and resistance to chipping.
  • Meet ASTM E1404 Specifications
  • High resistance to chemicals
  • Uniform wall thickness
  • Reinforced beaded rims to resist chipping
  • Dual metric scale graduations
  • Durable white enamel printing
  • Large white marking area
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)