MilliporeSigma
GLOBE-8454000
Globe Scientific
Globe Scientific Globe Glass Filter Flasks
Flask, Filter, Globe Glass, 4000mL, Dual Graduations, ASTM E1406, 1/Box
Shipped & Sold By Globe Scientific LLC
A Trusted Marketplace Seller
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
Globe Scientific
General description
Globe Glass™ heavy duty Erlenmeyer flasks meet ASTM E1406 specifications. Thick, heavy-duty walls improve mechanical strength and allow use in filtering applications not requiring side tubulation. Globe Glass heavy duty Erlenmeyer flasks are designed with reinforced, beaded rims for strength and resistance to chipping.
  • Meet ASTM E1406, Type I Specifications
  • Made exclusively from ASTM E438, Type I, Class A, 3.3 borosilicate glass
  • GlassGuard™ packaging system protects against transit damage
  • Thicker walls protect against damage
  • High resistance to chemicals
  • Uniform wall thickness
  • Reinforced beaded rims to resist chipping
  • Dual metric scale graduations
  • Durable white enamel printing
  • Large white marking area
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)