manufacturer/tradename
Golden West Diagnostics, LLC
General description
Source: Human Defibrinated Plasma
Country of Origin: United States
Filtration: 0.1 µm
Analytes: Reference specification sheet
Storage Temp: -20°C
Definition: Reference specification sheet
Still not finding the right product?
Explore all of our products under Golden West Diagnostics Mass Spect Gold Human Serum
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.