manufacturer/tradename
Golden West Diagnostics, LLC
General description
Source: Human
Country of Origin: United States
Analytes: Reference specification sheet
Storage Temp: -20°C
Definition: Defined as -15°C to -30°C
Still not finding the right product?
Explore all of our products under Golden West Diagnostics Human Urine
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.