Skip to Content
MilliporeSigma

GWDG-SP1040-1L

Golden West Diagnostics, LLC

Golden West Diagnostics Human Serum

1L, Charcoal Treated from Defibrinated Plasma


A Trusted Marketplace Seller

Synonym(s):

Processed serum, charcoal treated

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


manufacturer/tradename

Golden West Diagnostics, LLC

General description

Source: Human Defibrinated Plasma

Country of Origin: United States

Filtration: 0.22 µm

Analytes: Reference specification sheet

Storage Temp: -20°C

Definition: Reference specification sheet



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library