Skip to Content
MilliporeSigma

GDFH9BC79C21

Goodfellow

Silver Disk

Silver Disk,Formula: AgPercentage Purity: 99.97%Thickness: 0.02mmDiameter: 6mmCAS Number: 7440-22-4UOM Code: 669-447-40Legacy Code: AG000190Distributor Code: GF66944740

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
0
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Shipping Temperature

Ambient

Purity

99.97%

Storage temp.

Ambient

Country of Origin

US

Physical Form

Solid

Melting Point

961.9 C

Density

10.5g/cm³



Still not finding the right product?

Explore all of our products under Silver Disk




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library