MilliporeSigma
GROW-ISDW-2000
GROWCELLS
Growcells Water, Optimized for CO2 Incubators
EA, 2L bottle
Shipped & Sold By GROWCELLS
A Trusted Marketplace Seller
Synonym(s):
Co2 Incubator Water, Sterilized water
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
Growcells, Inc.
General description
Water for CO2 Incubators is double distilled, 0.1um filter sterilized, has a pH of 7-9 with a conductivity of 1-20 microSiemens (Resistivity of 50K-1 M ohm) and is terminal sterilized. The water is free of all contaminants and exceeds the specifications set by CO2 Incubator manufacturers.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)