MilliporeSigma
GROW-MBLE-3270
GROWCELLS
Growcells Terrific Broth Filter & Steam Sterilized (47.6 g/L)
EA, 1L bottle
Shipped & Sold By GROWCELLS
A Trusted Marketplace Seller
Synonym(s):
Terrific Broth, TB Broth
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
Growcells, Inc.
General description
TB broth, also known as Terrific Broth, is a nutrient-rich medium used in microbiology and molecular biology for growing bacteria, particularly E. coli. It is often employed in protein expression experiments and other applications where high cell densities are required.
Filter and Steam Sterilized.
12g Tryptone, 24g Yeast Extract, 9.4g Dibasic Potassium Phosphate, 2.2g Monobasic Potassium Phosphate, Adjusted to 7.0 +/- 0.2
  • High-Density Cultures: TB broth is often used when high cell densities are needed, such as for large-scale protein expression or biomass production.
  • Protein Expression: It supports the growth of E. coli and enhances the production of recombinant proteins, making it useful in various biotechnology and research applications
  • General Growth: Suitable for general bacterial cultivation, where enhanced growth and yield are desired.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)