MilliporeSigma
GROW-NUPW-1000
GROWCELLS
Growcells Water, Molecular Biology Grade. Nuclease Free
EA, 1L bottle
Shipped & Sold By GROWCELLS
A Trusted Marketplace Seller
Synonym(s):
Nuclease Free water, Sterile Water, General Molecular Biology Grade Water
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
Growcells, Inc.
General description
Certified Nuclease Free water is ideal for all applications that can be sensitive to residual bi-products of DEPC breakdown. Ideal for transformation, transfection, oocyte injection and all other research applications. Water is sterile filtered and steam sterilized inside final packaging to ensure sterility.
Growcells provides the highest quality of Nuclease Free water for critical applications. Water is purified, tested and certified to be devoid of any activiy in DNAse, RNAse, and Protease assays. Available in convenient volumes and packaging to fit all applications requiring biologically inactive water. Polycarbonate bottle packaging is re-usable and autoclavable.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)