MilliporeSigma
GROW-PCPW-1000
GROWCELLS
Growcells Water, PCR Grade, Certified, Protease, Nuclease & Endotoxin Free
Case (100), 1.8ml unit
Shipped & Sold By GROWCELLS
A Trusted Marketplace Seller
Synonym(s):
PCR Water, PCR grade nuclease free water
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
Growcells, Inc.
General description
This product is ideal for use in any PCR or RT-PCR application. Tested for prokaryotic as well as eukaryotic genomic DNA contamination that may cause false-positive signals in RT-PCR. Water is nuclease free and sterile filtered into conveniently sized vials for PCR use.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)