MilliporeSigma
GROW-SDW-0500
GROWCELLS
Growcells Water, Sterile, Distilled and Autoclaved
EA, 500ml bottle
Shipped & Sold By GROWCELLS
A Trusted Marketplace Seller
Synonym(s):
Sterile Water, Distilled Water, Cell culture Quality
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
Growcells, Inc.
General description
Used the preparation of cell culture media and laboratory reagents. Cell culture quality and molecular biological grade.
Growcells provides the highest quality of Nuclease Free water for critical applications. Water is purified, tested and certified to be devoid of any activiy in DNAse, RNAse, and Protease assays. Available in convenient volumes and packaging to fit all applications requiring biologically inactive water. Polycarbonate bottle packaging is re-usable and autoclavable.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)