MilliporeSigma
GROW-UPW-1000
GROWCELLS
Growcells Water, DEPC Treated, RNase/DNase, Nuclease Free, Ultrapure
EA, 1L bottle
Shipped & Sold By GROWCELLS
A Trusted Marketplace Seller
Synonym(s):
DEPC Treated Water, Ultrapure Water, Rnase free, Dnase free
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
Growcells, Inc.
General description
DEPC treatment is an effective way to eliminate RNAse activity. Our DEPC-treated water is made using the least amount of DEPC that is still effective at destroying enzymatic activitiy in RNases and other proteins, making it ideal for RNA extraction and all molecular biology applications. Water is autoclaved both before and after packaging to ensure sterility and to complete inactivation of byproducts of DEPC. CDI and UV treated.
Growcells provides the highest quality of Nuclease Free water for critical applications. Water is purified, tested and certified to be devoid of any activiy in DNAse, RNAse, and Protease assays. Available in convenient volumes and packaging to fit all applications requiring biologically inactive water. Polycarbonate bottle packaging is re-usable and autoclavable.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)