Skip to Content
MilliporeSigma

DW-30L528D

Haier Biomedical

Haier Biomedical -30C Biomedical Freezer

Upright cabinet, Capacity (L/Cu.Ft) 531/18.8, Power Supply (V/Hz) 115/60, Cooling Performance -30, Exterior Dimension (W*D*H)(in) 37*33.3*73.9

Shipped & Sold By Haier Biomedical

A Trusted Marketplace Seller

Synonym(s):

Biomedical Freezer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


weight

441 lb

H × W × D

2044 mm x 997 mm x 927 mm

manufacturer/tradename

Haier Biomedical USA

General description

Biomedical -30℃ Freezer (Direct Cooling)

Haier Biomedical -30℃ biomedical freezer with smart control and energy-saving hydrocarbon refrigeration system for safe and reliable product storage.

Applications: Biomedical and life science laboratories within blood banks, hospitals, disease control centres and research institutes as well as electronics and chemical industries. Used for cryopreservation of plasma, biological samples and other products and cold tests for components and materials. Temperature range -10°C to -30°C.



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library