Skip to Content
MilliporeSigma

DW-86L728BPST

Haier Biomedical

Haier Biomedical TwinCool Frequency Conversion ULT Freezer

25.7 cu ft, Temperature Range: -40~-86, Power Supply: 100~230/50/60 (V/Hz), Capacity (L/Cu.Ft): 728/25.7, Exterior Dimensions (W*D*H)(in): 41.2*39.3*78.0

Shipped & Sold By Haier Biomedical

A Trusted Marketplace Seller

Synonym(s):

Freezer, -80, ultra-low

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


weight

849 lb

H × W × D

2150 mm x 1090 mm x 1050 mm

manufacturer/tradename

Haier Biomedical USA

General description

Suitable for the storage of biological samples such as viruses, blood cells, bacteria and tissue, as well as electronics and other special materials.

The freezer combines frequency conversion compressors, hydrocarbon refrigerants (HC) and dual refrigeration system technology to deliver optimal sample security, energy-efficiency, and sustainability.

The product is used within hospitals, disease control, life science and medical research, bioengineering, pharma and other enterprize laboratories.

Product Features
  • Dual refrigeration system, double safety and optimal reliability: Double independent hydrocarbon refrigeration system design, each system can maintain -80ºC ensuring the safety of stored samples.
  • World-leading Energy Saving Refrigeration Technology: Intelligent frequency conversion technology and HC refrigeration system delivers a 50% reduction in energy usage when compared to traditional HFC refrigeration systems.
  • Environmental Protection: Environmentally friendly hydrocarbon refrigerant and foam material LBA.
  • Optional IoT System: The optional IoT monitors the operating status of the equipment in real time. Equipped with multiple alarm functions and a self-diagnostics system to identify and warns users to ensure sample safety.
  • Large loading capacity: Maximum Capacity: 600 boxes (2 inch boxes with 10*10 configuration) 60,000 samples can be stored.
  • Low Noise Design: Optimised systems and noise-reducing cabinet design lower noise out to 53dB for a quieter working experience.
  • Adjustable Sample Loading Try (Optional): The adjustable height sample load tray provides users with a convenient place to hold samples while opening and accessing storage compartments.
  • Intelligent Control: 10.1-inch high-performance touch screen, sensitive touch operation. Users can check the real-time operating status via IoT.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library