MilliporeSigma
DW-86L729BPT
Haier Biomedical
Haier Biomedical Smart Frequency Conversion ULT Freezer
25.7 cu ft, Power Supply: 100~230/50/60 (V/Hz), Touch Screen LCD Display, 4 Inner Doors, 3 Shelves, Exterior Dimension(W*D*H)(in) 41.2*39.3*78
Shipped & Sold By Haier Biomedical
A Trusted Marketplace Seller
Synonym(s):
Freezer, -80, ultra-low
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
weight
849 lb
H × W × D
2150 mm x 1090 mm x 1050 mm
manufacturer/tradename
Haier Biomedical USA
General description
Applicable for products and samples which require strict storage conditions such as viruses, pathogens, red blood cells, white blood cells, skin, bones, bacteria, semen, biological products, electronics and special materials.
Suitable for long term storage applications and compliant with typical storage requirements found in hospitals, disease control and prevention centres, scientific research institutions biomedical engineering institutes, agriculture/ fishery companies as well as the electronics and chemical industry.
Product Features
  • Safe and secure: Standard equipped with key lock, padlock and with optional fingerprint lock and electromagnetic lock, providing multiple safeguards for sample safety.
  • Low Noise design reducing noise down to 53dB: Special noise-reduction design plus super silent compressor technology and energy-saving fan, considerably lowers noise level.
  • Super energy efficient with three environmentally friendly innovations: Ultra-low power consumption, down to less than 10kWh/day, ensuring a world leading energy saving performance.
  • Optimized insulation: Double foaming for both inner and outer doors and five-layer sealing design and optimized super-thick VIP thermal insulation technology, extends temperature holdover time during power failure and increases insulation efficiency by 20%.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)