Skip to Content

JWPH3249CBDB

J & W Pharmlab LLC

(S)-Tomoxetine hydrochloride

Synonym(s):

(3S)-N-methyl-3-(2-methylphenoxy)-3-phenyl-1-propanamine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C17H22O1N1Cl1
Molecular Weight:
291.82
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C17H21NO.ClH/c1-14-8-6-7-11-16(14)19-17(12-13-18-2)15-9-4-3-5-10-15;/h3-11,17-18H,12-13H2,1-2H3;1H/t17-;/m0./s1

Inchi Key

LUCXVPAZUDVVBT-LMOVPXPDSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library