Skip to Content
MilliporeSigma

MAN182108496

Manchester Organics

1,1,1-Trifluoroacetone

Synonym(s):

1,1,1-trifluoroacetone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
112.05
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C3H3F3O/c1-2(7)3(4,5)6/h1H3

Inchi Key

FHUDAMLDXFJHJE-UHFFFAOYSA-N

Purity

97%

Country of Origin

GB



Still not finding the right product?

Explore all of our products under 1,1,1-Trifluoroacetone




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library