MilliporeSigma
All Photos(1)

Documents

03-0076-02

Millipore

SMC® Detection Antibody Labeling Kit

Sign Into View Organizational & Contract Pricing

Synonym(s):
SMC® Detection Labeling Kit, Single Molecule Counting Detection Labeling Kit
eCl@ss:
32161000

manufacturer/tradename

SMC®

Quality Level

technique(s)

ELISA: suitable
single molecule counting: suitable

General description

Take advantage of SMC® proprietary digital Single Molecule Counting technology while developing your own application-specific immunoassays. This kit provides everything you need to build your own high performing SMC® immunoassay. It includes reagents and consumables to label up to 1mg of antibody to detection antibody tag.

Application

Develop your own detection antibodies and move beyond the traditional ELISA with SMC® Technology.
Label up to 1mg of antibody to detection antibody tag

Components

Detection label
1X Buffer 1
1X Buffer 2
10X Buffer
3 Filter Tube Ultrafree
Ultra 4 Filter Tube

Legal Information

SMC is a registered trademark of Singulex

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service