MilliporeSigma

1.00810

SAFC

Trichloroacetic acid

crystalline, EMPROVE® ESSENTIAL, Ph. Eur.

Synonym(s):
Trichloroacetic acid
Empirical Formula (Hill Notation):
C2HCl3O2
CAS Number:
Molecular Weight:
163.39
MDL number:
EC Index Number:
200-927-2

Agency

Ph. Eur.

Quality Level

vapor pressure

1 hPa ( 20 °C)

product line

EMPROVE® ESSENTIAL

Assay

≥99.0-100.5% (alkalimetric)

form

crystalline
solid

autoignition temp.

711 °C

pH

<1 (20 °C, 50 g/L in H2O)

mp

54-56 °C

transition temp

flash point >110 °C

solubility

water: 1300 g/L at 20 °C

density

1.63 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤0.01%

application(s)

liquid formulation
pharma/biopharma processes
pharmaceutical
semi-solid formulation

storage temp.

15-25°C

InChI

1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

YNJBWRMUSHSURL-UHFFFAOYSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Trichloroacetic acid is typically used in topical formulations.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

The substance 1.00810 has no REACH registration because the only supported use is the use in medicinal products for human or veterinary use or in food or feedingstuffs according to article 2 of the REACH Regulation (EC) No 1907/2006.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Slide 1 of 2

1 of 2

Trichloroacetic acid suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), &#8805;99%

Sigma-Aldrich

T8657

Trichloroacetic acid

2-Thiobarbituric acid &#8805;98%

Sigma-Aldrich

T5500

2-Thiobarbituric acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service