MilliporeSigma
All Photos(2)

Documents

1.00978

Supelco

Isoamyl alcohol

(mixture of isomers) for determination of fat acc. to Gerber

Sign Into View Organizational & Contract Pricing

Synonym(s):
Isoamyl alcohol, iso-Pentyl alcohol, 3-Methyl-1-butanol, Iso-Amylalcohol
Empirical Formula (Hill Notation):
C5H12O
CAS Number:
Molecular Weight:
88.15
MDL number:
EC Index Number:
250-378-8
Web Name Override:
iso-Pentyl alcohol
Web Name Suffix Override:
mixture of isomers, for use in fat content analysis

vapor pressure

3.1 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

340 °C

expl. lim.

1.2-8 % (v/v)

bp

131 °C/1013 hPa

mp

-117 °C

transition temp

flash point 43 °C

density

0.81 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C5H12O/c1-2-3-4-5-6/h6H,2-5H2,1H3

InChI key

AMQJEAYHLZJPGS-UHFFFAOYSA-N

General description

Isoamyl alcohol is a clear, colorless alcohol. It is one of several isomers of amyl alcohol. It is commonly used as a solvent in various applications, such as inks, coatings, and pharmaceutical preparations.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Slide 1 of 2

1 of 2

3-Methyl-1-butanol anhydrous, ≥99%

Sigma-Aldrich

309435

3-Methyl-1-butanol

Phenol solution Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology

Sigma-Aldrich

P4557

Phenol solution

Pentanol isomer synthesis in engineered microorganisms
Cann AF,et al.
Applied Microbiology and Biotechnology, 85, 893-899 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service