MilliporeSigma

1.03814

SAFC

Iron(III) chloride hexahydrate

EMPROVE® ESSENTIAL, Ph. Eur., JPE

Synonym(s):
Ferric chloride hexahydrate, Iron trichloride, Ferric chloride hexahydrate
Linear Formula:
FeCl3 · 6H2O
CAS Number:
Molecular Weight:
270.30
MDL number:
EC Index Number:
231-729-4

Agency

JPE
Ph. Eur.

Quality Level

vapor pressure

1 mmHg ( 194 °C)

product line

EMPROVE® ESSENTIAL

form

solid

potency

316 mg/kg LD50, oral (Rat)

pH

1.8 (25 °C, 10 g/L in H2O)

bp

280-285 °C (lit.)

mp

37 °C (lit.)

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

15-25°C

SMILES string

O.O.O.O.O.O.Cl[Fe](Cl)Cl

InChI

1S/3ClH.Fe.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

NQXWGWZJXJUMQB-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
1.039434494431232
form

solid

form

solid

form

crystals or chunks

form

solid

pH

1.8 (25 °C, 10 g/L in H2O)

pH

1.8 (25 °C, 10 g/L in H2O)

pH

-

pH

-

mp

37 °C (lit.)

mp

37 °C (lit.)

mp

37 °C (lit.)

mp

37 °C (lit.)

Quality Level

500

Quality Level

300

Quality Level

-

Quality Level

-

agency

JPE

agency

reag. Ph. Eur.

agency

-

agency

USP/NF, reag. Ph. Eur.

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Iron(III) chloride hexahydrate shows adstringent and hemostatic properties. Application includes veterinary medicine.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Slide 1 of 5

1 of 5

Iron(III) chloride anhydrous for synthesis

Sigma-Aldrich

8.03945

Iron(III) chloride

Iron(III) chloride anhydrous, powder, ≥99.99% trace metals basis

Sigma-Aldrich

451649

Iron(III) chloride

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service