MilliporeSigma

1.06643

SAFC

Sodium sulfate

anhydrous, EMPROVE® ESSENTIAL, Ph. Eur., BP, USP, E 514

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
MDL number:
E Number:
E514
EC Index Number:
231-820-9

Agency

BP
Ph. Eur.
USP

Quality Level

product line

EMPROVE® ESSENTIAL

form

solid

pH

5.2-8.0 (20 °C, 50 g/L in H2O)

mp

884 °C (lit.)

solubility

200 g/L

density

2.68 g/mL at 25 °C (lit.)

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sodium sulfate anhydrous is the anhydrous, sodium salt form of sulfuric acid, and can be used as buffer and pH modifier or mineral salt.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Slide 1 of 8

1 of 8

Sodium sulfate decahydrate crystalline, EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JPE, USP

SAFC

1.06642

Sodium sulfate decahydrate

Sodium sulfate anhydrous, granular, free-flowing, Redi-Dri™, ACS reagent, ≥99%

Sigma-Aldrich

798592

Sodium sulfate

Sodium sulfate suitable for HPLC, LiChropur™, 99.0-101.0% (T)

Supelco

80948

Sodium sulfate

Sodium sulfate anhydrous for synthesis

Sigma-Aldrich

8.22286

Sodium sulfate

Sodium sulfate BioUltra, anhydrous, ≥99.0% (T)

Sigma-Aldrich

71959

Sodium sulfate

Sodium sulfate puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)

Sigma-Aldrich

13464

Sodium sulfate

Sodium Molybdate, Dihydrate Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Supelco

SX0650

Sodium Molybdate, Dihydrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service