Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.07983

SAFC

Sulfur

Sulfur for external use Ph Eur,BP. CAS 7704-34-9, molar mass 32.06 g/mol.

Sign Into View Organizational & Contract Pricing

Synonym(s):
Sulfur
Empirical Formula (Hill Notation):
S
CAS Number:
Molecular Weight:
32.07
MDL number:
UNSPSC Code:
12141912
EC Index Number:
231-722-6

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

form

solid

autoignition temp.

235 °C (Dust)

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

bp

444 °C/1013 hPa

mp

113-119 °C

transition temp

flash point 160 °C

density

1.96-2.07 g/cm3 at 20 °C

bulk density

400‑500 kg/m3

application(s)

pharma/biopharma processes

storage temp.

2-30°C

InChI

1S/H2S/h1H2

Inchi Key

RWSOTUBLDIXVET-UHFFFAOYSA-N

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Sulfur can be used in chemical synthesis or pharmaceutical processes.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 6

1 of 6

Sigma-Aldrich

Sigma-Aldrich

24245

Oil of turpentine

Potassium dichromate ACS reagent, &#8805;99.0%

Sigma-Aldrich

207802

Potassium dichromate

Methyl alcohol United States Pharmacopeia (USP) Reference Standard

USP

1424109

Methyl alcohol

Sodium dodecyl sulfate 92.5-100.5% based on total alkyl sulfate content basis

Sigma-Aldrich

L5750

Sodium dodecyl sulfate

Polysorbate 80 tested according to Ph. Eur.

Sigma-Aldrich

59924

Polysorbate 80

Lead oxide powder (red), 1-2&#160;&#956;m, 99%

Sigma-Aldrich

241547

Lead oxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service