MilliporeSigma
All Photos(3)

1.37042

SAFC

TRI-SODIUM CITRATE DIHYDRATE

crystalline, EMPROVE® EXPERT, Ph. Eur., BP, JP, USP, ACS

Synonym(s):
Sodium citrate tribasic dihydrate, Trisodium Citrate, Sodium Citrate, Dihydrate, Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate
Linear Formula:
HOC(COONa)(CH2COONa)2 · 2H2O
CAS Number:
Molecular Weight:
294.10
Beilstein:
6104939
MDL number:
EC Index Number:
200-675-3

grade

ACS reagent

Quality Level

Agency

BP
JP
Ph. Eur.
USP

description

EMPROVE® EXPERT Ph Eur,BP,JP,USP,ACS

product line

EMPROVE® EXPERT

form

crystalline
solid

pH

7.5-9.0 (25 °C, 50 g/L in H2O)

mp

150 °C (anhydrous substance)
>300 °C (lit.)

application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical

storage temp.

2-30°C

SMILES string

O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3Na.2H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;2*1H2/q;3*+1;;/p-3

InChI key

NLJMYIDDQXHKNR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

1 of 4

This Item
1.064321.064461.06447
TRI-SODIUM CITRATE DIHYDRATE crystalline, EMPROVE® EXPERT, Ph. Eur., BP, JP, USP, ACS

SAFC

1.37042

TRI-SODIUM CITRATE DIHYDRATE

tri-Sodium citrate dihydrate crystalline, EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, USP, E 331

SAFC

1.06432

tri-Sodium citrate dihydrate

tri-Sodium citrate dihydrate powder, EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, USP, E 331

SAFC

1.06446

tri-Sodium citrate dihydrate

tri-Sodium citrate dihydrate EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JP, USP, ACS, E 331

SAFC

1.06447

tri-Sodium citrate dihydrate

grade

ACS reagent

grade

-

grade

-

grade

-

agency

BP, JP, Ph. Eur., USP

agency

BP, JP, Ph. Eur., USP

agency

BP, JP, Ph. Eur., USP

agency

BP, ChP, JP, Ph. Eur., USP

product line

EMPROVE® EXPERT

product line

EMPROVE® ESSENTIAL

product line

EMPROVE® ESSENTIAL

product line

EMPROVE® ESSENTIAL

form

crystalline, solid

form

crystalline, solid

form

powder, solid

form

solid

pH

7.5-9.0 (25 °C, 50 g/L in H2O)

pH

7.5-9.0 (25 °C, 50 g/L in H2O)

pH

7.5-9.0 (25 °C, 50 g/L in H2O)

pH

7.5-9.0 (25 °C, 50 g/L in H2O)

mp

150 °C (anhydrous substance), >300 °C (lit.)

mp

150 °C (anhydrous substance), >300 °C (lit.)

mp

150 °C (anhydrous substance), >300 °C (lit.)

mp

150 °C (anhydrous substance), >300 °C (lit.)

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

tri-Sodium citrate dihydrate is typically used as a citrate buffer system component for biomolecule downstream chromatography steps or final liquid formulation.
Due to its low microbial and endotoxin limits, tri-Sodium citrate dihydrate cryst. Emprove® Expert is especially suitable for high-risk applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

(enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service