Skip to Content
MilliporeSigma

1.37228

T7 RNA Polymerase

greener alternative

EMPROVE® EXPERT

Synonym(s):

Polymerase, T7, T7 RNAP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Specific activity:
≥50.000 units/mL
Assay:
≥95% (SDS-PAGE)
Biological source:
bacteriophage T7
Recombinant:
expressed in E. coli
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bacteriophage T7

Quality Segment

recombinant

expressed in E. coli

grade

GMP, Halal, Kosher

product line

EMPROVE® EXPERT

assay

≥95% (SDS-PAGE)

form

aqueous solution

specific activity

≥50.000 units/mL

mol wt

99 kDa

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

in vitro transcription (IVT): suitable

suitability

suitable for (In‑vitro transcription (IVT))

application(s)

pharma/biopharma processes
pharmaceutical

foreign activity

Low endotoxin
Low bioburden
DNase: none detected
RNase: none detected
Nickase: none detected

greener alternative category

storage temp.

-10 to -25°C

General description

mRNA technologies represent a revolutionary approach for prophylactic vaccines and for therapeutics, including cancer treatments and immunotherapies. As global interest in mRNA manufacturing grows and regulatory expectations become more stringent, manufacturers require raw materials suitable for pharmaceutical use with high quality and purity to enable successful mRNA production.

In response, we have developed the high-purity enzyme Emprove® Expert portfolio, compliant with IPEC-PQG GMP standards, to ensure batch-to-batch consistency in your mRNA manufacturing processes. These critical raw materials are accompanied by comprehensive Emprove® Dossiers, providing essential regulatory support to facilitate compliance and accelerate time to market in the biopharmaceutical industry.

As part of our SAFC® portfolio, these high-quality raw materials are designed to meet the specific needs of pharma and biopharma production.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product falls under category 12 PAP and aligns with the Green Chemistry Principle of "Designing Safer Chemicals". It is a preparation with defined composition that replaces Triton X-100 with Tween-20, addressing concerns of Triton X-100 as an endocrine disruptor, delivering improved safety.

Application

T7 RNA polymerase is the core enzyme used in in vitro transcription (IVT) workflows for the production of messenger RNA (mRNA). It catalyzes the synthesis of RNA from a DNA template containing a T7 promoter sequence. Its high specificity and robust transcription efficiency make it the standard choice for manufacturing mRNA therapeutics and vaccines.

Features and Benefits

Our IVT enzyme Emprove® Expert IVT portfolio is designed to align with best-practice IVT methods for mRNA manufacturing and provides all purity requirements for reliable production of mRNA therapeutics and vaccines:
  • High purity (≥95%)
  • Low endotoxin and bioburden
  • DNase and RNase-free
Designed with therapeutic mRNA manufacturing in mind, our range of Emprove® Expert IVT enzymes provide all the quality attributes required for therapeutic mRNA raw materials, with validated quality control testing and manufacturing processes, certifications for AOF, Kosher, and Halal, and an extensive range of quality parameters provided in the COA.

Packaging

137228.0001 / 1 mL / plastic vial
137228.0010 / 10 mL / plastic bottle
137228.0050 / 50 mL / plastic bottle

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under T7 RNA Polymerase


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library