MilliporeSigma
All Photos(1)

Documents

1.46145

Millipore

GSP Agar

selective for bacteria: Pseudomonas spp., selective for bacteria: Aeromonas spp., pack of 20 unit

Sign Into View Organizational & Contract Pricing

Synonym(s):
Pseudomonas Aeromonas Selective Agar

sterility

sterile; autoclaved

Quality Level

form

powder

packaging

pack of 20 unit

technique(s)

microbiological culture: suitable

final pH

7.2±0.2 (25 °C)

application(s)

food and beverages

storage temp.

15-25°C

suitability

selective for bacteria: Aeromonas spp.
selective for bacteria: Pseudomonas spp.

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
50875856271.10235
vibrant-m

1.46145

GSP Agar

vibrant-m

50875

GSP Agar

vibrant-m

85627

SPS Agar

vibrant-m

1.10235

SPS agar

sterility

sterile; autoclaved

sterility

non-sterile

sterility

non-sterile

sterility

-

form

powder

form

powder

form

powder

form

medium granules (dehydrated (DCM))

suitability

selective for bacteria: Aeromonas spp., selective for bacteria: Pseudomonas spp.

suitability

Aeromonas spp., selective and differential for Pseudomonas spp.

suitability

selective and differential for Clostridium spp.

suitability

Clostridium perfringens, Clostridium spp.

application(s)

food and beverages

application(s)

bioburden testing
environmental
food and beverages
water monitoring, microbiology

application(s)

bioburden testing
environmental
food and beverages, microbiology

application(s)

food and beverages, microbiology

storage temp.

15-25°C

storage temp.

-

storage temp.

-

storage temp.

15-25°C

General description

GSP Agar is designed for selective detection and isolation of Pseudomonas and Aeromonas spp. in food, wastewater, and equipment samples from food industry.
Accompanying microflora cannot metabolize glutamate and starch present in the media which enhances the selectivity of the medium. Selective growth on the medium is indicated by color change of phenol red to yellow for Aeromonas spp. Pseudomonas spp. growth will not indicate any color change from red.

Application

GSP Agar is used for selective detection and isolation of Pseudomonas and Aeromonas spp. in test samples from food industry.

Features and Benefits

Prepared Plates are clear and red.
Pseudomonas spp.: Large, diameter 2-3mm, blue-violet, surrounded by a red violet zone.
Aeromonas spp.: Large, diameter 2-3mm, yellow, surrounded by a yellow zone.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service