MilliporeSigma
All Photos(4)

Documents

ABN1641

Sigma-Aldrich

Anti-APP585C

serum, from rabbit

Sign Into View Organizational & Contract Pricing

Synonym(s):
Amyloid beta A4 protein, ABPP, APPI, APP, Alzheimer disease amyloid protein, Amyloid precursor protein, Beta-amyloid precursor protein, Cerebral vascular amyloid peptide, CVAP, PreA4, Protease nexin-II, PN-II
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

species reactivity (predicted by homology)

canine (based on 100% sequence homology), chimpanzee (based on 100% sequence homology), rabbit (based on 100% sequence homology), rat (based on 100% sequence homology), porcine (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... APP(351)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
MABN2295MABN2297MABN2287
Anti-APP585C serum, from rabbit

ABN1641

Anti-APP585C

Anti-APP Antibody, clone 7H6 clone 7H6, from rat

MABN2297

Anti-APP Antibody, clone 7H6

Anti-APP Antibody, clone 1D1 clone 1D1, from rat

MABN2287

Anti-APP Antibody, clone 1D1

biological source

rabbit

biological source

rat

biological source

rat

biological source

rat

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

clone

polyclonal

clone

2E9, monoclonal

clone

7H6, monoclonal

clone

1D1, monoclonal

species reactivity

human, mouse

species reactivity

mouse, human

species reactivity

human

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

technique(s)

immunofluorescence: suitable, immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

ELISA: suitable, flow cytometry: suitable, immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

ELISA: suitable, flow cytometry: suitable, immunocytochemistry: suitable, immunohistochemistry: suitable (paraffin), western blot: suitable

Still not finding the right product?  

Give our Product Selector Tool a try.

General description

Amyloid beta A4 protein (UniProt: P05067; also known as ABPP, APPI. APP, APP, Alzheimer disease amyloid protein, Amyloid precursor protein, Beta-amyloid precursor protein, Cerebral vascular amyloid peptide, CVAP, PreA4, Protease nexin-II, PN-II) is encoded by the APP (also known as A4, AD1) gene (Gene ID: 351) in human. Deposition of Amyloid beta (Ab) is an early event in the pathogenesis of Alzheimer s disease (AD). Ab peptides originate from the proteolytic cleavage of the amyloid precursor protein (APP). APPs are cell surface protein that are internalized via clathrin-coated pits. During maturation, the immature APP (N-glycosylated in the endoplasmic reticulum) moves to the Golgi complex where complete maturation occurs (O-glycosylated and sulfated). After alpha-secretase cleavage, soluble APP is released into the extracellular space and the C-terminal is internalized to endosomes and lysosomes. The beta-secretase cleaves APP between residues Met671 and Asp672 and yields sAPP beta and C99. Following the beta-secretase cleavage, a second cleavage occurs at the C-terminus of Ab peptide that releases Ab from C99. This cleavage occurs in the vicinity of residue 712 of the C-terminus. The gamma-secretase can cleave the C-terminal region at either Val711 or Ile713 to produce the shorter Ab peptide (Ab1-40) or the longer Ab peptide (Ab1-42). Ab1-42 occurs more frequently and forms fibrillar aggregates far more readily than the Ab1-40 peptide. Beta-amyloid peptides are lipophilic metal chelators with metal-reducing activity. Bind transient metals such as copper, zinc and iron. In vitro, can reduce Cu2+ and Fe3+ to Cu+ and Fe2+, respectively. Abeta42 is a more effective reductant than beta-amyloid 40. From a physiological point of view, it functions on the surface of neurons relevant to neurite growth, neuronal adhesion and axonogenesis. It is also involved in cell mobility and transcription regulation through protein-protein interactions and can promote transcription activation through binding to APBB1-KAT5.

Specificity

This polyclonal antibody detects amyloid precursor protein in human Alzheimer′s disese brain. It targets an epitope within 9 amino acids in the vicintiy of APP585 in the C-terminal region.

Immunogen

A linear peptide corresponding to 9 amino acids from the C-terminal region of human Amyloid beta A4 protein.
Epitope: extracellular domain

Application

Anti-APP585C, Cat. No. ABN1641, is a highly specific rabbit polyclonal antibody that targets Amyloid beta A4 protein and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis (WB): A 1:1000 Dilution of this antibody detected Amyloid beta A4 protein in 5XFAD and 5XFAD/AEP-/- mouse brain lysate.
Western Blotting Analysis (WB): A Dilution of this antibody detected Amyloid beta A4 protein in PMID 26549211 .
Immunohistochemistry Analysis (IHC): A Dilution of this antibody detected Amyloid beta A4 protein in PMID 26549211 .
Immunohistochemistry Analysis (IHC): A 1:200 Dilution of this antibody detected Amyloid beta A4 protein in 5XFAD familial AD transgenic mouse brain.

Quality

Evaluated by Immunohistochemistry in human Alzheimer′s diseased human brain tissue.

Immunohistochemistry Analysis: A 1:200 dilution of this antibody detected APP585C in Alzheimer′s diseased human brain tissue.

Target description

10 kDa calculated.

Physical form

Rabbit polyclonal antiserum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service