MilliporeSigma
All Photos(1)

Documents

CFSP001750

Millipore

C083 Cellulose Fiber Sample Pad Roll

Lateral Flow, Assay Development, lateral flow immunoassays, lateral flow assay device, Immunoassay IVDs

Sign Into View Organizational & Contract Pricing

eCl@ss:
32031602

Quality Level

packaging

pack of 1 roll

manufacturer/tradename

SureWick®

roll W × L

17 mm × 50 m

width

17 mm

shipped in

ambient

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
103882MHWP037A0MAWP025AC
vibrant-m

103882

Cell Culture Media 4CHO Sample Kit

manufacturer/tradename

SureWick®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

packaging

pack of 1 roll

packaging

-

packaging

-

packaging

-

roll W × L

17 mm × 50 m

roll W × L

-

roll W × L

-

roll W × L

-

width

17 mm

width

-

width

-

width

-

Application

Lateral Flow, Assay Development, lateral flow immunoassays, lateral flow assay device, Immunoassay IVDs

Legal Information

SUREWICK is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service