MilliporeSigma
All Photos(1)

Documents

EQ70XXPKT1H

Milli-Q

Milli-Q® EQ 7008/16 H Purification Cartridge Kit

Designed for use with Milli-Q® EQ 7008/16 systems for the production of RO pure (Type 3) and ultrapure (Type 1) water

Sign Into View Organizational & Contract Pricing

material

carbon block
carbon filter (activated)
cartridge (polyphosphate beads)
filter (Pleated)
filter (soda lime)
media (IQnano ion exchange)
media (Jetpore® mixed-bed ion exchange)

Quality Level

feature

Kit contains IPAK Gard® H Pretreatment Pack, IPAK Meta®, Polishing CartridgeIPAK Quanta® Polishing Cartridge and a Tank Vent Filter.

manufacturer/tradename

Milli-Q®

compatibility

for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016

shipped in

ambient

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EQ70XXPKT1IX700XPKT1HIX70XXPKT1
manufacturer/tradename

Milli-Q®

manufacturer/tradename

Milli-Q®

manufacturer/tradename

Milli-Q® IX

manufacturer/tradename

Milli-Q® IX

material

carbon block

material

carbon block, carbon filter (activated), filter (Pleated), filter (soda lime), media (IQnano ion exchange), media (Jetpore® mixed-bed ion exchange)

material

-

material

-

feature

Kit contains IPAK Gard® H Pretreatment Pack, IPAK Meta®, Polishing CartridgeIPAK Quanta® Polishing Cartridge and a Tank Vent Filter.

feature

Kit contains IPAK Gard® Pretreatment Pack, IPAK Meta®, Polishing CartridgeIPAK Quanta® Polishing Cartridge and a Tank Vent Filter.

feature

-

feature

-

shipped in

ambient

shipped in

ambient

shipped in

-

shipped in

-

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

General description

This water purification kit includes the following:

  • IPAK Gard® H Pretreatment Pack (for hard water) efficiently removes colloids, particles and free chlorine from tap water to protect the reverse osmosis (RO) membrane and improve system performance. Polyphosphate beads provided added protection against scaling of the RO membrane. Recommended for feed water hardness (CaCO3) > 300 ppm.
  • IPAK Meta® and IPAK Quanta® polishing cartridges contain innovative Jetpore® mixed-bed ion-exchange resin and IQnano ion-exchange media, which achieve ion removal down to trace levels. In addition, high-grade synthetic activated carbon targets trace organic contaminants.
  • The Tank Vent Filter, redesigned for seamless integration, provides improved protection against airborne contaminants.

This kit is designed for use with Milli-Q® EQ 7008/16 systems for the production of RO pure (Type 3) and ultrapure (Type 1) water.

Application

For optimal protection of the RO membrane, trace ion and organics removal, and advanced protection against airborne contaminants.

Features and Benefits

IPAK Gard® H Pretreatment Pack (for hard water) contains:
  • Activated carbon to protect against free chlorine.
  • Polyphosphate beads to protect against scaling and precipitates.
  • Depth filter to protect against particulates and colloids.

Tank Vent Filter contains:
  • Activated carbon to remove volatile organics.
  • Soda lime to remove CO2.
  • Membrane filter to prevent particulates and bacteria from entering the tank.

IPAK Meta® cartridge contains:
  • Jetpore® mixed-bed ion-exchange resin and IQnano ion-exchange media to remove ions down to trace levels.


IPAK Quanta® Cartridge contains:
  • Innovative IQnano ion-exchange media to remove ions down to trace levels.
  • High-grade synthetic activated carbon targets trace organic contaminants.

Legal Information

IPAK GARD is a registered trademark of Merck KGaA, Darmstadt, Germany
IPAK META is a registered trademark of Merck KGaA, Darmstadt, Germany
IPAK QUANTA is a registered trademark of Merck KGaA, Darmstadt, Germany
IQNANO is a trademark of Merck KGaA, Darmstadt, Germany
JETPORE is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Related Content

Always have confidence in your science with Milli-Q® ultrapure water. Discover our expanded system offer to find the lab water solution that fully satisfies your lab’s needs and constraints.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service