MilliporeSigma
All Photos(1)

Documents

HF18002NSS

Millipore

HI-Flow Plus HF18002

new

Lateral Flow, Assay Development, lateral flow immunoassays, lateral flow assay device, Immunoassay IVDs

eCl@ss:
32031602

Quality Level

packaging

sheet of 5 unit

parameter

180 sec/4 cm capillary flow rate

backing thickness

2 mil (50 μm)

filter W × L

30.5 cm × 25.0 cm

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
HF180MC5PKHF180MC100HF18004NSS
Quality Level

400

Quality Level

400

Quality Level

400

Quality Level

400

packaging

sheet of 5 unit

packaging

pkg of 5 ea

packaging

pkg of 100 ea

packaging

sheet of 5 unit

backing thickness

2 mil (50 μm)

backing thickness

-

backing thickness

-

backing thickness

4 mil (100 μm)

filter W × L

30.5 cm × 25.0 cm

filter W × L

6 cm × 30.1 cm

filter W × L

6 cm × 30.1 cm

filter W × L

30.5 cm × 25.0 cm

General description

Sheets of Hi-Flow plus Cellulose Ester membranes, 2 mil backed with nominal capillary flow time of 180 seconds/4 cm

Legal Information

HI-Flow is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service