MilliporeSigma
All Photos(2)

Documents

IX700XPKT1H

Milli-Q

Milli-Q® IX Purification Cartridge Kit

for use with Milli-Q® IX 7003/05, Protects the RO membrane and preserves stored pure (Type 2) water quality, fed with hardwater

Sign Into View Organizational & Contract Pricing

Quality Level

composition

Soda lime (for CO2 removal)

packaging

pkg of 1 kit

manufacturer/tradename

Milli-Q® IX

compatibility

for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
IX700XPKT1IX70XXPKT1EQ70XXPKT1
manufacturer/tradename

Milli-Q® IX

manufacturer/tradename

Milli-Q® IX

manufacturer/tradename

Milli-Q® IX

manufacturer/tradename

Milli-Q®

compatibility

for use with Milli-Q® IX 7003, for use with Milli-Q® IX 7005

compatibility

for use with Milli-Q® IX 7003, for use with Milli-Q® IX 7005

compatibility

for use with Milli-Q® IX 7010, for use with Milli-Q® IX 7015

compatibility

for use with Milli-Q® EQ 7008, for use with Milli-Q® EQ 7016

composition

Soda lime (for CO2 removal)

composition

Soda lime (for CO2 removal)

composition

Soda lime (for CO2 removal)

composition

-

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

General description

This water purification cartridge kit includes the following:
  • IPAK Gard® H 3-5 Pretreatment Pack (for hard water) efficiently removes colloids, particles, free chlorine, organics and minerals for reverse osmosis (RO) membrane protection and improved system performance.
  • Tank Vent filter, redesigned for seamless integration, provides improved protection against airborne contaminants.

Application

For optimal protection of the RO membrane and advanced protection against airborne contaminants.

Features and Benefits

IPAK Gard® H 3-5 Pretreatment Pack (for hard water) contains:
  • Activated carbon to protect against free chlorine, and organics.
  • Polyphosphate beads to protect against scaling and precipitates.
  • Depth filter to protect against particulates and colloids.
  • 63% of the parts suppliers for IPAK Gard® cartridge included in this kit are ISO 14001 environmental management system-certified.

Tank Vent Filter contains:
  • Activated carbon to remove volatile organics.
  • Soda lime to remove CO2.
  • Membrane filter to prevent particulates and bacteria from entering the tank.

Legal Information

IPAK GARD is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service