MilliporeSigma
All Photos(2)

Documents

MABE1669

Sigma-Aldrich

Anti-Cas9 Antibody, NT Antibody, clone 8C1-F10

clone 8C1-F10, from mouse

Sign Into View Organizational & Contract Pricing

Synonym(s):
CRISPR-associated endonuclease Cas9/Csn1, SpyCas9
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8C1-F10, monoclonal

species reactivity

Streptococcus pyogenes

species reactivity (predicted by homology)

all

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
MABE1668MABS1291ABC1490
clone

8C1-F10, monoclonal

clone

6G12-H11, monoclonal

clone

10C11-A12, monoclonal

clone

polyclonal

biological source

mouse

biological source

mouse

biological source

mouse

biological source

rabbit

species reactivity

Streptococcus pyogenes

species reactivity

Streptococcus pyogenes

species reactivity

bacteria

species reactivity

Streptococcus pyogenes

technique(s)

immunocytochemistry: suitable, western blot: suitable

technique(s)

immunocytochemistry: suitable, western blot: suitable

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

shipped in

ambient

shipped in

ambient

shipped in

wet ice

shipped in

ambient

Still not finding the right product?  

Give our Product Selector Tool a try.

General description

CRISPR-associated endonuclease Cas9/Csn1 (UniProt Q99ZW2; also known as SpyCas9) is encoded by the Streptococcus pyogenes serotype M1 gene Cas9 (also known as csn1) gene (Gene ID 901176). Clustered regularly interspaced short palindromic repeats (CRISPRs) are DNA loci containing short repetitions of base sequences complementary to antecedent mobile elements and target invading nucleic acids. Each repetition is followed by short segments of spacer DNA from previous exposures to a virus. Cas9 is an essential endonuclease in Streptococcus pyogenes serotype M1′s CRISPR immune system that confers resistance to foreign genetic elements such as plasmids and phages. CRISPR clusters are transcribed and processed into CRISPR RNA (crRNA). In type II CRISPR systems, correct processing of pre-crRNA requires a trans-encoded small RNA (tracrRNA), endogenous ribonuclease 3 (rnc), and Cas9. Cas9/crRNA/tracrRNA endonucleolytically cleaves linear or circular dsDNA target complementary to the spacer. The target strand not complementary to crRNA is first cut endonucleolytically, and then trimmed 3′-5′ exonucleolytically.

Specificity

Clone 8C1-F10 targets an epitope within Cas9 N-terminal region outside the active site RuvC-I.

Immunogen

His-tagged recombinant Cas9 N-terminal fragment excluding the active site RuvC-I.

Application

Anti-Cas9, NT, clone 8C1-F10 , Cat. No. MABE1669, is a highly specific mouse monoclonal antibody that targets CRISPR-associated endonuclease Cas9/Csn1 and has been tested in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected exogenously expressed full-length nuclease-dead Cas9 in transfected NIH/3T3 cells fixed with 4% paraformaldehyde and permeabilized by 0.3% Triton X-100.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in NIH3T3 cells.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected exogenously expressed full-length nuclease-dead Cas9 in 10 µg of lysate from transfected NIH/3T3 cells.

Target description

~160 kDa observed. 158.4 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service