MilliporeSigma
All Photos(2)

Documents

MC0HC23CL3

Millipore

Millistak+® Depth Filter in μPOD format

C0HC, cellulose fiber, filtration area 0.0023 m2, hydrophilic

Sign Into View Organizational & Contract Pricing

Synonym(s):
Millistak+Depth Filter in μPod format, C0HC media series, 23 cm2 surface area, Luer fitting connection
eCl@ss:
32031690
NACRES:
NB.24

material

cellulose fiber
filter aid (inorganic)
glass fiber-reinforced polypropylene

Quality Level

description

Perfusion bioreactor fluid

feature

hydrophilic

manufacturer/tradename

Millistak+®

parameter

1.0 bar max. differential pressure (15 psid) at 40 °C (Reverse)
2.1 bar max. differential pressure (30 psid) at 40 °C (Forward)

H

6.6 cm (2.6 in.)

L

8.9 cm (3.5 in.)

filtration area

0.0023 m2

impurities

<0.25 EU/mL (LAL test, Aqueous extraction)

gravimetric extractables

420-750 mg/ft2

input

sample type liquid

conductivity

3.64 – 10.5 μS/cm (product water; post autoclave (1 cycle of 60 minutes at 123 °C) and pure water flush of 10 liter per ft²(100 L/m²) of media surface area)

filter grade

C0HC

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
MCE2023CL3MCE4023CL3MCE4523CL3
material

cellulose fiber , glass fiber-reinforced polypropylene , filter aid (inorganic)

material

cellulose , glass fiber-reinforced polypropylene

material

cellulose , glass fiber-reinforced polypropylene

material

cellulose , glass fiber-reinforced polypropylene

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic

description

Perfusion bioreactor fluid

description

For clarification of serum, plasma, vaccines, cell culture or other fluids.

description

For clarification of serum, plasma, vaccines, cell culture or other fluids.

description

For clarification of serum, plasma, vaccines, cell culture or other fluids.

manufacturer/tradename

Millistak+®

manufacturer/tradename

Millistak+®

manufacturer/tradename

Millistak+®

manufacturer/tradename

Millistak+®

parameter

1.0 bar max. differential pressure (15 psid) at 40 °C (Reverse)

parameter

1.0 bar max. differential pressure (15 psid) at 40 °C (Reverse), 2.1 bar max. differential pressure (30 psid) at 40 °C (Forward)

parameter

1.0 bar max. differential pressure (15 psid) at 40 °C (Reverse), 2.1 bar max. differential pressure (30 psid) at 40 °C (Forward)

parameter

1.0 bar max. differential pressure (15 psid) at 40 °C (Reverse), 2.1 bar max. differential pressure (30 psid) at 40 °C (Forward)

General description

The Millistak+® depth filter media is offered in a scalable, disposable format providing greater flexibility because of its unique modular design. The Millistak+® MC0HC23CL3 comes in a μPOD format featuring a 6.6 cm (H) x 8.9 cm (L) and a filtration area of 0.0023 m2.

This C0HC media series delivers optimal performance through a gradient density matri, ideal for primary clarification applications. Millistak+® Pod filters are available in three distinct series of media grades in order to meet your specific needs - DE, CE and HC.

The Millistak+® Pod system is ideal for a wide variety of primary and secondary clarification applications, including cell cultures, yeast and E. coli lysates post centrifuge, E. coli refolds, media, vaccines, plasma proteins and sera.

Easy to Use

With the compact, modular design of our Millistak+® new Pod system, you can increase productivity and shorten cycle times.

Installation and setup of the Pod system is simple and straightforward. The unique design of the disposable adapters and disposable manifolds makes it easy to connect the Pod filters to rest of the unit operations in the process. The self-contained and disposable nature of the system protects operators from exposure to biohazards and eliminates maintenance as well as cleaning validation requirements.

Application

Perfusion bioreactor fluid

Features and Benefits

  • Low hold-up volume for greater product yield
  • Broad range of media types offered in single and multilayer products
  • Millistak+® HC dual-action media improves prefiltration and compresses clarification
  • Flexible, modular format offers scalability from 5 to 12,000 liters or more
  • Patented disposable design eliminates need for housing, CIP or cleaning validation
  • Self-contained Pod filters protect operators from exposure to biohazards
  • Robust construction is easy to use and set up
  • Smaller footprint facilitates use in tight spaces

Specifications

Filter Grade: C0HC

Preparation Note

Sterilization Method
2 autoclave cycles of 60 minutes at 123 °C.

Analysis Note

Gravimetric Extractables: per 24-hour static soak in pure water; (type 1 DI water), post autoclave (1 cycle of 60 minutes at 123 °C) and pure water flush of 10 liters per ft2 (100 L/m2) of media surface area

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service